Learn More
2,4,6-Trimethoxybenzonitril, 98 %, Thermo Scientific Chemicals
Catalog Number p-7027072
Click to view available options
Menge:
1 g
5 g
25 g
2571-54-2
C10H11NO3
193.202
MFCD00001787
GBRHJUMDNWLSCT-UHFFFAOYSA-N
75731
2,4,6-trimethoxybenzonitril
COC1=CC(=C(C(=C1)OC)C#N)OC
2571-54-2
C10H11NO3
193.202
MFCD00001787
GBRHJUMDNWLSCT-UHFFFAOYSA-N
75731
2,4,6-trimethoxybenzonitril
COC1=CC(=C(C(=C1)OC)C#N)OC
2,4,6-Trimethoxybenzonitrile is used as pharmaceutical intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Anwendungen
2,4,6-Trimethoxybenzonitril wird als pharmazeutisches Zwischenprodukt verwendet.
Löslichkeit
Unlöslich in Wasser.
Hinweise
Kühl lagern. Behälter an einem trockenen und gut belüfteten Ort dicht verschlossen halten. Inkompatible Materialien sind Oxidationsmittel.
TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 2,4,6-Trimethoxybenzonitrile
Schmelzpunkt 140°C to 143°C
Geruch Characteristic
Menge 1 g
Synonym benzonitrile, 2,4,6-trimethoxy, 2,4,6-trimethoxy-benzonitrile, 2,4,6-trimethoxybenzenecarbonitrile, acmc-20aoeu, 2,4,6-trimethoxy benzonitrile, benzonitrile,2,4,6-trimethoxy, 2,4,6-trimethoxybenzonitrile
Löslichkeitsinformationen Insoluble in water.
Formelmasse 193.2
Reinheit (%) 98%