missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichlor-5-Fluorbenzoesäure, 97 %, Thermo Scientific™

Catalog Number p-7026201
Click to view available options
Menge:
1 g
5 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
86522-89-6
C7H2Cl2FO2
207.99
MFCD00075281
KZCWJHUTTSVCRO-UHFFFAOYSA-M
2,4-dichloro-5-fluoro-benzoic acid, 2,4-dichloro-5-fluoro benzoic acid, benzoic acid, 2,4-dichloro-5-fluoro, pubchem1338, maybridge1_002392, acmc-209qa7, ksc448a5p, rarechem al be 1219, timtec-bb sbb003459, 2 4-dichloro-5-fluorobenzoic acid
688138
2,4-dichloro-5-fluorobenzoate
[O-]C(=O)C1=CC(F)=C(Cl)C=C1Cl
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
86522-89-6
C7H2Cl2FO2
207.99
MFCD00075281
KZCWJHUTTSVCRO-UHFFFAOYSA-M
2,4-dichloro-5-fluoro-benzoic acid, 2,4-dichloro-5-fluoro benzoic acid, benzoic acid, 2,4-dichloro-5-fluoro, pubchem1338, maybridge1_002392, acmc-209qa7, ksc448a5p, rarechem al be 1219, timtec-bb sbb003459, 2 4-dichloro-5-fluorobenzoic acid
688138
2,4-dichloro-5-fluorobenzoate
[O-]C(=O)C1=CC(F)=C(Cl)C=C1Cl

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 2,4-Dichloro-5-fluorobenzoic acid
Schmelzpunkt 140°C to 142°C
Menge 1 g
Formelmasse 209.01
Reinheit (%) 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'