missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dihydroxybenzhydrazid, 95 %, Thermo Scientific Chemicals

Catalog Number p-7045241
Click to view available options
Menge:
5 g
25 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
13221-86-8
C7H8N2O3
168.15
MFCD00017053
GOPUHTXVZGIOHN-UHFFFAOYSA-N
2,4-dihydroxybenzhydrazide, 2,4-dihydroxybenzoic acid hydrazide, benzoic acid, 2,4-dihydroxy-, hydrazide, 2,4-dihydroxybenzoic hydrazide, 2,4-bis oxidanyl benzohydrazide, 2,4-dihydroxybenzoic acid, hydrazide, acmc-1cf1y, 4-???benzene-1,3-diol, 2,4-dihydroxybenzoylhydrazine, 2,4-dihydroxy benzoic acid hydrazide
166785
2,4-Dihydroxybenzohydrazid
NNC(=O)C1=CC=C(O)C=C1O
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
13221-86-8
C7H8N2O3
168.15
MFCD00017053
GOPUHTXVZGIOHN-UHFFFAOYSA-N
2,4-dihydroxybenzhydrazide, 2,4-dihydroxybenzoic acid hydrazide, benzoic acid, 2,4-dihydroxy-, hydrazide, 2,4-dihydroxybenzoic hydrazide, 2,4-bis oxidanyl benzohydrazide, 2,4-dihydroxybenzoic acid, hydrazide, acmc-1cf1y, 4-???benzene-1,3-diol, 2,4-dihydroxybenzoylhydrazine, 2,4-dihydroxy benzoic acid hydrazide
166785
2,4-Dihydroxybenzohydrazid
NNC(=O)C1=CC=C(O)C=C1O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 2,4-Dihydroxybenzhydrazide
Schmelzpunkt 240°C to 242°C
Menge 5 g
Beilstein 2641320
Formelmasse 168.15
Reinheit (%) 95%

RUO – Nur für Forschungszwecke

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'