missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dimethoxypyrimidin, 98+ %, Thermo Scientific Chemicals

Catalog Number p-7028467
Click to view available options
Menge:
1 g
5 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
3551-55-1
C6H8N2O2
140.142
MFCD00056028
KEVRHVMWBKFGLO-UHFFFAOYSA-N
pyrimidine, 2,4-dimethoxy, unii-980k6dh82b, dimethoxypyrimidine, zlchem 892, pubchem7104, acmc-1adzt, 2,4-dimethoxy-pyrimidine, ksc911k2l, 2,4-dimethoxypyrimidine, kevrhvmwbkfglo-uhfffaoysa
137976
2,4-Dimethoxypyrimidin
COC1=NC(=NC=C1)OC
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
3551-55-1
C6H8N2O2
140.142
MFCD00056028
KEVRHVMWBKFGLO-UHFFFAOYSA-N
pyrimidine, 2,4-dimethoxy, unii-980k6dh82b, dimethoxypyrimidine, zlchem 892, pubchem7104, acmc-1adzt, 2,4-dimethoxy-pyrimidine, ksc911k2l, 2,4-dimethoxypyrimidine, kevrhvmwbkfglo-uhfffaoysa
137976
2,4-Dimethoxypyrimidin
COC1=NC(=NC=C1)OC

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 2,4-Dimethoxypyrimidine
Schmelzpunkt 17°C
Dichte 1.1501
Siedepunkt 204°C to 205°C
Flammpunkt 93°C (199°F)
Brechungsindex 1.502
Menge 1 g
Formelmasse 140.14
Reinheit (%) ≥98%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'