missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dinitrobenzylbromid, TRC

Artikelnummer. 30371557
Change view
Click to view available options
Menge:
100 mg
missing translation for 'unitSize'
100 Milligramm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Menge unitSize
30371557 100 mg 100 Milligramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 30371557 missing translation for 'mfr' Toronto Research Chemicals missing translation for 'supplierNo' TRCD680470100MG

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

This item is not returnable. View return policy

Hochreine organische Moleküle und analytische Standards, strategisch weltweit geliefert, um Innovation und wirtschaftlichen Erfolg zu ermöglichen.

TRUSTED_SUSTAINABILITY

Specifications

Chemischer Name oder Material 2,4-Dinitrobenzyl Bromide
CAS 3013-38-5
Summenformel C7 H5 Br N2 O4
Menge 100 mg
Synonym 1-(Bromomethyl)-2,4-dinitrobenzene (ACI), Toluene, α-bromo-2,4-dinitro- (6CI, 7CI, 8CI), 2,4-Dinitrobenzyl bromide, o,p-Dinitrobenzyl bromide
InChI-Formel InChI=1S/C7H5BrN2O4/c8-4-5-1-2-6(9(11)12)3-7(5)10(13)14/h1-3H,4H2
SMILES [O-][N+](=O)c1ccc(CBr)c(c1)[N+](=O)[O-]
IUPAC-Name 1-(Brommethyl)-2,4-dinitrobenzol
Molekulargewicht (g/mol) 261.03
Formelmasse 259.943
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.