missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Methoxyphenoxy)Propylamin, 99 %

Catalog Number p-7033581
Click to view available options
Menge:
250 mg
1 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
93750-30-2
C10H15NO2
181.235
MFCD02684116
HIWWBKQBOHWJIM-UHFFFAOYSA-N
2-4-methoxyphenoxy propylamine, 2-4-methoxyphenoxy propan-1-amine, 1-1-aminopropan-2-yl oxy-4-methoxybenzene, 2-4-methoxy-phenoxy-propylamine, acmc-20aop0, pha-q01-0, 4-1-aminoprop-2-yl oxy anisole, 2-4-methoxyphenoxy-1-propanamine, alpha-aminomethyl-4-methoxyphenetole
5199614
2-(4-methoxyphenoxy)propan-1-amin
CC(CN)OC1=CC=C(C=C1)OC
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
93750-30-2
C10H15NO2
181.235
MFCD02684116
HIWWBKQBOHWJIM-UHFFFAOYSA-N
2-4-methoxyphenoxy propylamine, 2-4-methoxyphenoxy propan-1-amine, 1-1-aminopropan-2-yl oxy-4-methoxybenzene, 2-4-methoxy-phenoxy-propylamine, acmc-20aop0, pha-q01-0, 4-1-aminoprop-2-yl oxy anisole, 2-4-methoxyphenoxy-1-propanamine, alpha-aminomethyl-4-methoxyphenetole
5199614
2-(4-methoxyphenoxy)propan-1-amin
CC(CN)OC1=CC=C(C=C1)OC

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 2-(4-Methoxyphenoxy)propylamine
Menge 250 mg
UN-Nummer UN2735
Empfindlichkeit Air Sensitive
Formelmasse 181.24
Reinheit (%) 99%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'