Learn More
2-(4-Methylperhydro-1,4-diazepin-1-yl)benzoesäure-hydrochloridhemihydrat, 95 %, Thermo Scientific™
Catalog Number p-3522422
Click to view available options
Menge:
1 g
5 g
Verpackung:
Bernsteinfarbene Glasflasche
921938-77-4
C13H19ClN2O2
270.757
MFCD09065004
UCYNEWGETFFMQY-UHFFFAOYSA-N
2-4-methyl-1,4-diazepan-1-yl benzoic acid hydrochloride, 2-4-methyl-1,4-diazepan-1-yl benzoic acid hcl, 2-4-methylhomopiperazin-1-yl benzoic acid hydrochloride hemihydrate, 2-4-methyl-1,4-diazepan-1-yl benzoic acid-hydrogen chloride 1/1, 2-4-methylperhydro-1,4-diazepin-1-yl benzoic acid hydrochloride 0.5 hydrate, benzoicacid, 2-hexahydro-4-methyl-1h-1,4-diazepin-1-yl-, hydrochloride 1:1, benzoicacid,2-hexahydro-4-methyl-1h-1,4-diazepin-1-yl-,hydrochloride 1:1
24229621
2-(4-methyl-1,4-diazepan-1-yl)benzoesäure;hydrochlorid
CN1CCCN(CC1)C2=CC=CC=C2C(=O)O.Cl
921938-77-4
C13H19ClN2O2
270.757
MFCD09065004
UCYNEWGETFFMQY-UHFFFAOYSA-N
2-4-methyl-1,4-diazepan-1-yl benzoic acid hydrochloride, 2-4-methyl-1,4-diazepan-1-yl benzoic acid hcl, 2-4-methylhomopiperazin-1-yl benzoic acid hydrochloride hemihydrate, 2-4-methyl-1,4-diazepan-1-yl benzoic acid-hydrogen chloride 1/1, 2-4-methylperhydro-1,4-diazepin-1-yl benzoic acid hydrochloride 0.5 hydrate, benzoicacid, 2-hexahydro-4-methyl-1h-1,4-diazepin-1-yl-, hydrochloride 1:1, benzoicacid,2-hexahydro-4-methyl-1h-1,4-diazepin-1-yl-,hydrochloride 1:1
24229621
2-(4-methyl-1,4-diazepan-1-yl)benzoesäure;hydrochlorid
CN1CCCN(CC1)C2=CC=CC=C2C(=O)O.Cl
Chemischer Name oder Material 2-(4-Methylperhydro-1, 4-diazepin-1-yl)benzoic acid hydrochloride hemihydrate
CAS Min. % 94.5
CAS Max. % 100.0
Prozentgehaltsbereich 95%
Verpackung Bernsteinfarbene Glasflasche
Menge 1 g
Formelmasse 279.77
Reinheit (%) 95%