missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Pyridyl)-imidazol-4,5-dithiocarboxamid, 97 %, Thermo Scientific™

Catalog Number 15441536
Click to view available options
Menge:
250 mg
1 g
5 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
1347815-03-5
C10H9N5S2
263.337
MFCD20265330
DZNBSARSJBFIQR-UHFFFAOYSA-N
2-4-pyridyl imidazole-4,5-dithiocarboxamide, 2-pyridin-4-yl-1h-imidazole-4,5-dicarbothioamide
73995943
2-Pyridin-4-yl-1H-Imidazol-4,5-Dicarbothioamid
C1=CN=CC=C1C2=NC(=C(N2)C(=S)N)C(=S)N
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
1347815-03-5
C10H9N5S2
263.337
MFCD20265330
DZNBSARSJBFIQR-UHFFFAOYSA-N
2-4-pyridyl imidazole-4,5-dithiocarboxamide, 2-pyridin-4-yl-1h-imidazole-4,5-dicarbothioamide
73995943
2-Pyridin-4-yl-1H-Imidazol-4,5-Dicarbothioamid
C1=CN=CC=C1C2=NC(=C(N2)C(=S)N)C(=S)N

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 2-(4-Pyridyl)imidazole-4,5-dithiocarboxamide
Menge 250 mg
Formelmasse 263.33
Reinheit (%) 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'