missing translation for 'onlineSavingsMsg'
Learn More

2,5-Difluorbenzenboronsäure, 96 %, Thermo Scientific Chemicals

Catalog Number p-7027395
Click to view available options
Menge:
1 g
5 g
25 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
193353-34-3
C6H5BF2O2
157.911
MFCD01863171
KTOJGSDLJNUAEP-UHFFFAOYSA-N
2,5-difluorophenyl boronic acid, 2,5-difluorobenzeneboronic acid, 2,5-difluorophenyl boranediol, boronic acid, 2,5-difluorophenyl, 2,5-difluorophenylboronicacid, 2,5-fluorobenzene boronic acid, pubchem1820, acmc-1c1am, ksc174i6r, 2,5-difluoro phenylboronic acid
2734335
(2,5-difluorphenyl)boronsäure
B(C1=C(C=CC(=C1)F)F)(O)O
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
193353-34-3
C6H5BF2O2
157.911
MFCD01863171
KTOJGSDLJNUAEP-UHFFFAOYSA-N
2,5-difluorophenyl boronic acid, 2,5-difluorobenzeneboronic acid, 2,5-difluorophenyl boranediol, boronic acid, 2,5-difluorophenyl, 2,5-difluorophenylboronicacid, 2,5-fluorobenzene boronic acid, pubchem1820, acmc-1c1am, ksc174i6r, 2,5-difluoro phenylboronic acid
2734335
(2,5-difluorphenyl)boronsäure
B(C1=C(C=CC(=C1)F)F)(O)O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 2,5-Difluorobenzeneboronic acid
Menge 1 g
Beilstein 8833254
Formelmasse 157.91
Reinheit (%) 96%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'