missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dimethyl-1,3,4-thiadiazol, ≥ 99 %, Thermo Scientific™

Catalog Number p-7023555
Click to view available options
Menge:
5 g
25 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
27464-82-0
C4H6N2S
114.166
MFCD00020722
JXQGICFGPUAVLJ-UHFFFAOYSA-N
96922
2,5-Dimethyl-1,3,4-Thiadiazol
CC1=NN=C(S1)C
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
27464-82-0
C4H6N2S
114.166
MFCD00020722
JXQGICFGPUAVLJ-UHFFFAOYSA-N
96922
2,5-Dimethyl-1,3,4-Thiadiazol
CC1=NN=C(S1)C

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 2,5-Dimethyl-1,3,4-thiadiazole
Schmelzpunkt 61°C to 65°C
Menge 5 g
Synonym 2,5-dimethylthiadiazole, 1,3,4-thiadiazole, 2,5-dimethyl, dimethyl-1,3,4-thiadiazole, unii-frm8gdw64z, frm8gdw64z, pubchem23972, acmc-1cr4f, 1,4-thiadiazole, 2,5-dimethyl, 1,3,4-thiadiazole,2,5-dimethyl, 2,5-dimethyl-1,3,4-thiadiazole
Formelmasse 114.17
Reinheit (%) ≥99%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'