missing translation for 'onlineSavingsMsg'
Learn More

2,6-Difluor-3-Methoxyphenylessigsäure, 97 %, Thermo Scientific™

Catalog Number p-7029283
Click to view available options
Menge:
250 mg
1 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
886498-65-3
C9H8F2O3
202.157
MFCD04115908
KIUMRKHFRAIFFF-UHFFFAOYSA-N
2,6-difluoro-3-methoxyphenylacetic acid, 2-2,6-difluoro-3-methoxyphenyl acetic acid, 2,6-difluoro-3-methoxyphenyl acetic acid, benzeneacetic acid,2,6-difluoro-3-methoxy
3816042
2-(2,6-difluor-3-methoxyphenyl)essigsäure
COC1=C(C(=C(C=C1)F)CC(=O)O)F
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
886498-65-3
C9H8F2O3
202.157
MFCD04115908
KIUMRKHFRAIFFF-UHFFFAOYSA-N
2,6-difluoro-3-methoxyphenylacetic acid, 2-2,6-difluoro-3-methoxyphenyl acetic acid, 2,6-difluoro-3-methoxyphenyl acetic acid, benzeneacetic acid,2,6-difluoro-3-methoxy
3816042
2-(2,6-difluor-3-methoxyphenyl)essigsäure
COC1=C(C(=C(C=C1)F)CC(=O)O)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 2,6-Difluoro-3-methoxyphenylacetic acid
Menge 250 mg
Formelmasse 202.16
Reinheit (%) 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'