missing translation for 'onlineSavingsMsg'
Learn More

2,6-Difluor-4-(Methoxycarbonyl)Benzenboronsäure Pinacolester, 96 %

Catalog Number p-7039780
Click to view available options
Menge:
250 mg
1 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
1218791-32-2
C14H17BF2O4
298.093
MFCD12546491
CGADBVGBEFYTMK-UHFFFAOYSA-N
methyl 3,5-difluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzoate, 2,6-difluoro-4-methoxycarbonyl phenylboronic acid pinacol ester, 2,6-difluoro-4-methoxycarbonyl phenylboronic acid, pinacol ester, 2,6-difluoro-4-methoxycarbonyl benzeneboronic acid pinacol ester, 2,6-difluoro-4-methoxycarbonyl phenyl boronic acid pinacol ester, 2,6-difluoro-4-methoxycarbonyl phenylboronic acid,pinacol ester, methyl 3,5-difluoro-4-tetramethyl-1,3,2-dioxaborolan-2-yl benzoate, methyl3,5-difluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzoate
46739549
methyl-3,5-difluor-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoat
B1(OC(C(O1)(C)C)(C)C)C2=C(C=C(C=C2F)C(=O)OC)F
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
1218791-32-2
C14H17BF2O4
298.093
MFCD12546491
CGADBVGBEFYTMK-UHFFFAOYSA-N
methyl 3,5-difluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzoate, 2,6-difluoro-4-methoxycarbonyl phenylboronic acid pinacol ester, 2,6-difluoro-4-methoxycarbonyl phenylboronic acid, pinacol ester, 2,6-difluoro-4-methoxycarbonyl benzeneboronic acid pinacol ester, 2,6-difluoro-4-methoxycarbonyl phenyl boronic acid pinacol ester, 2,6-difluoro-4-methoxycarbonyl phenylboronic acid,pinacol ester, methyl 3,5-difluoro-4-tetramethyl-1,3,2-dioxaborolan-2-yl benzoate, methyl3,5-difluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzoate
46739549
methyl-3,5-difluor-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoat
B1(OC(C(O1)(C)C)(C)C)C2=C(C=C(C=C2F)C(=O)OC)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 2,6-Difluoro-4-(methoxycarbonyl)benzeneboronic acid pinacol ester
Menge 250 mg
Formelmasse 298.09
Reinheit (%) 96%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'