missing translation for 'onlineSavingsMsg'
Learn More

2,6-Difluoranisol, 98+%, Thermo Scientific™

Catalog Number p-7027188
Click to view available options
Menge:
1 g
5 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
437-82-1
C7H6F2O
144.121
MFCD00142846
IOBWAHRFIPQEQL-UHFFFAOYSA-N
2,6-difluoroanisole, 1,3-difluoro-2-methoxy-benzene, benzene, 1,3-difluoro-2-methoxy, pubchem1968, 2,6-difluoro anisole, 2.6-difluoro anisole, acmc-1ajjj, ksc235o3h, iobwahrfipqeql-uhfffaoysa
2736897
1,3-Difluor-2-Methoxybenzol
COC1=C(C=CC=C1F)F
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
437-82-1
C7H6F2O
144.121
MFCD00142846
IOBWAHRFIPQEQL-UHFFFAOYSA-N
2,6-difluoroanisole, 1,3-difluoro-2-methoxy-benzene, benzene, 1,3-difluoro-2-methoxy, pubchem1968, 2,6-difluoro anisole, 2.6-difluoro anisole, acmc-1ajjj, ksc235o3h, iobwahrfipqeql-uhfffaoysa
2736897
1,3-Difluor-2-Methoxybenzol
COC1=C(C=CC=C1F)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 2,6-Difluoroanisole
Dichte 1.221
Siedepunkt 70°C to 72°C (56 mmHg)
Flammpunkt 42°C (108°F)
Menge 1 g
UN-Nummer UN1993
Formelmasse 144.12
Reinheit (%) ≥98%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'