missing translation for 'onlineSavingsMsg'
Learn More

2,6-Difluorbenzimid, 98+ %, Thermo Scientific™

Catalog Number p-7081586
Click to view available options
Menge:
25 g
100 g
500 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
18063-03-1
C7H5F2NO
157.12
MFCD00007972
AVRQBXVUUXHRMY-UHFFFAOYSA-N
87439
2,6-difluorbenzamid
C1=CC(=C(C(=C1)F)C(=O)N)F
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
18063-03-1
C7H5F2NO
157.12
MFCD00007972
AVRQBXVUUXHRMY-UHFFFAOYSA-N
87439
2,6-difluorbenzamid
C1=CC(=C(C(=C1)F)C(=O)N)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 2,6-Difluorobenzamide
Schmelzpunkt 145°C to 148°C
Menge 25 g
Beilstein 2047480
Synonym benzamide, 2,6-difluoro, 2,6-dfam, unii-sj2rn214mk, sj2rn214mk, 2,6-difluorobenzoylamide, 2,6 difluorobenzamide, 2.6-difluorobenzamide, pubchem10766, 2, 6 difluorobenzamide, 2,6 difluorobenzamide;
Formelmasse 157.12
Reinheit (%) ≥98%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'