missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4,5,6,7-tetrahydrobenzo[b]thiophen-3-carbonitril, 97 %, Thermo Scientific Chemicals

Catalog Number p-7031459
Click to view available options
Menge:
1 g
10 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
4651-91-6
C9H10N2S
178.253
MFCD00128278
ADHVMGAFAKSNOM-UHFFFAOYSA-N
2-amino-4,5,6,7-tetrahydrobenzo b thiophene-3-carbonitrile, 2-amino-3-cyano-4,5,6,7-tetrahydrobenzo b thiophene, 2-amino-4,5,6,7-tetrahydro-benzo b thiophene-3-carbonitrile, benzo b thiophene-3-carbonitrile, 2-amino-4,5,6,7-tetrahydro, 2-amino-4,5,6,7-tetrahydro-1-benzo b thiophene-3-carbonitrile, 2-amino-4,5,6,7-tetrahydrobenzo b thiophene-3-carbonitryle, cambridge id 5130181, aminotetrahydrobenzothiophenecarbonitrile
78382
2-Amino-4,5,6,7-Tetrahydro-1-Benzothiophen-3-Carbonitril
C1CCC2=C(C1)C(=C(S2)N)C#N
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
4651-91-6
C9H10N2S
178.253
MFCD00128278
ADHVMGAFAKSNOM-UHFFFAOYSA-N
2-amino-4,5,6,7-tetrahydrobenzo b thiophene-3-carbonitrile, 2-amino-3-cyano-4,5,6,7-tetrahydrobenzo b thiophene, 2-amino-4,5,6,7-tetrahydro-benzo b thiophene-3-carbonitrile, benzo b thiophene-3-carbonitrile, 2-amino-4,5,6,7-tetrahydro, 2-amino-4,5,6,7-tetrahydro-1-benzo b thiophene-3-carbonitrile, 2-amino-4,5,6,7-tetrahydrobenzo b thiophene-3-carbonitryle, cambridge id 5130181, aminotetrahydrobenzothiophenecarbonitrile
78382
2-Amino-4,5,6,7-Tetrahydro-1-Benzothiophen-3-Carbonitril
C1CCC2=C(C1)C(=C(S2)N)C#N

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 2-Amino-4,5,6,7-tetrahydrobenzo[b]thiophene-3-carbonitrile
Schmelzpunkt 141°C to 144°C
Menge 1 g
Formelmasse 178.26
Reinheit (%) 97%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'