missing translation for 'onlineSavingsMsg'
Learn More

2-Aminobenzylcyanid, Hydrochlorid, TRC

Artikelnummer. 30313909
Change view
Click to view available options
Menge:
2 g
Unit Size:
2 Gramm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Menge unitSize
30313909 2 g 2 Gramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30313909 Supplier Toronto Research Chemicals Supplier No. TRCA5969252G

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

This item is not returnable. View return policy

Hochreine organische Moleküle und analytische Standards, strategisch weltweit geliefert, um Innovation und wirtschaftlichen Erfolg zu ermöglichen.

TRUSTED_SUSTAINABILITY

Specifications

Chemischer Name oder Material 2-Aminobenzylcyanide, Hydrochloride
CAS 40943-70-2
Summenformel C8 H8 N2 . Cl H
Menge 2 g
Synonym Benzeneacetonitrile, 2-amino-, hydrochloride (1:1), Benzeneacetonitrile, 2-amino-, monohydrochloride (9CI), o-Aminophenylacetonitrile hydrochloride, 2-Aminobenzeneacetonitrile hydrochloride, (2-Aminophenyl)acetonitrile hydrochloride, 2-(Cyanomethyl)aniline hydrochloride, 2-Aminobenzyl cyanide hydrochloride, o-Aminobenzyl cyanide hydrochloride
InChI-Formel InChI=1S/C8H8N2.ClH/c9-6-5-7-3-1-2-4-8(7)10;/h1-4H,5,10H2;1H
SMILES Cl.Nc1ccccc1CC#N
IUPAC-Name 2-(2-Aminophenyl)acetonitril;hydrochlorid
Molekulargewicht (g/mol) 168.62
Formelmasse 168.0454
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.