Learn More
2-Aminobiphenyl-d9, TRC
Specifications
Specifications
| Chemischer Name oder Material | 2-Aminobiphenyl-d9 |
| CAS | 344298-97-1 |
| Summenformel | C12 D9 H2 N |
| Menge | 10 mg |
| Synonym | 2-Aminobiphenyl-d9 |
| InChI-Formel | InChI=1S/C12H11N/c13-12-9-5-4-8-11(12)10-6-2-1-3-7-10/h1-9H,13H2/i1D,2D,3D,4D,5D,6D,7D,8D,9D |
| SMILES | [2H]c1c([2H])c([2H])c(c([2H])c1[2H])c2c([2H])c([2H])c([2H])c([2H])c2N |
| IUPAC-Name | 2,3,4,5-Tetradeuterio-6-(2,3,4,5,6-Pentadeuteriophenyl)anilin |
| Molekulargewicht (g/mol) | 178.2779 |
| Formelmasse | 178.1456 |