Learn More
2-Benzyloxypyridin-3-Boronsäure-Pinacolester, 95 %, Thermo Scientific Chemicals
Catalog Number p-7029464
Click to view available options
Menge:
250 mg
1 g
1073371-81-9
C18H22BNO3
311.19
MFCD06798265
IFMSDMPTVPMGOP-UHFFFAOYSA-N
2-benzyloxy-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-benzyloxypyridine-3-boronic acid pinacol ester, acmc-20aajr, 2-benzyloxy pyridin-3-yl boronic acid pinacol ester, 2-benzyloxy-3-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl pyridine, 2-benzyloxy-3-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-benzoxy-3-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-pyridine, 2-phenylmethoxy-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-pyridine
44755162
2-(benzyloxy)-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
CC1(C)OB(OC1(C)C)C1=C(OCC2=CC=CC=C2)N=CC=C1
1073371-81-9
C18H22BNO3
311.19
MFCD06798265
IFMSDMPTVPMGOP-UHFFFAOYSA-N
2-benzyloxy-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-benzyloxypyridine-3-boronic acid pinacol ester, acmc-20aajr, 2-benzyloxy pyridin-3-yl boronic acid pinacol ester, 2-benzyloxy-3-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl pyridine, 2-benzyloxy-3-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-benzoxy-3-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-pyridine, 2-phenylmethoxy-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-pyridine
44755162
2-(benzyloxy)-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
CC1(C)OB(OC1(C)C)C1=C(OCC2=CC=CC=C2)N=CC=C1
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 2-Benzyloxypyridine-3-boronic acid pinacol ester
Menge 250 mg
Formelmasse 311.19
Reinheit (%) 95%