missing translation for 'onlineSavingsMsg'
Learn More

2-Brom-1,3-Dichlor-5-(Trifluoromethoxy)Benzol, 97 %, Thermo Scientific Chemicals

Catalog Number p-7031488
Click to view available options
Menge:
1 g
10 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
118754-55-5
C7H2BrCl2F3O
309.891
MFCD03412210
CAAJZKZQEJSRJO-UHFFFAOYSA-N
2-bromo-1,3-dichloro-5-trifluoromethoxy benzene, 2,6-dichloro-4-trifluoromethoxy bromobenzene, 4-bromo-3,5-dichloro-1-trifluoromethoxy benzene, benzene,2-bromo-1,3-dichloro-5-trifluoromethoxy, acmc-1c5si
18947696
2-Brom-1,3-dichlor-5-(trifluormethoxy)benzol
C1=C(C=C(C(=C1Cl)Br)Cl)OC(F)(F)F
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
118754-55-5
C7H2BrCl2F3O
309.891
MFCD03412210
CAAJZKZQEJSRJO-UHFFFAOYSA-N
2-bromo-1,3-dichloro-5-trifluoromethoxy benzene, 2,6-dichloro-4-trifluoromethoxy bromobenzene, 4-bromo-3,5-dichloro-1-trifluoromethoxy benzene, benzene,2-bromo-1,3-dichloro-5-trifluoromethoxy, acmc-1c5si
18947696
2-Brom-1,3-dichlor-5-(trifluormethoxy)benzol
C1=C(C=C(C(=C1Cl)Br)Cl)OC(F)(F)F

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 2-Bromo-1,3-dichloro-5-(trifluoromethoxy)benzene
Menge 1 g
Formelmasse 309.9
Reinheit (%) 97%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'