missing translation for 'onlineSavingsMsg'
Learn More

2-Brom-4,5-Dichlorimidazol, 98 %, Thermo Scientific Chemicals

Catalog Number 11482697
Click to view available options
Menge:
1 g
5 g
25 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
16076-27-0
C3HBrCl2N2
215.86
MFCD00067894
SULQOTGFPPFUMG-UHFFFAOYSA-N
2-bromo-4,5-dichloroimidazole, imidazole, 2-bromo-4,5-dichloro, 1h-imidazole, 2-bromo-4,5-dichloro, imidazole,5-dichloro, acmc-20ao2s, 1h-imidazole,5-dichloro, 5-23-04-00463 beilstein handbook reference, 2-bromo-4,5-dichloro-imidazole, 2-bromo-4,5-dichloro-3h-imidazole
99484
2-Brom-4,5-Dichlor-1H-Imidazol
ClC1=C(Cl)N=C(Br)N1
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
16076-27-0
C3HBrCl2N2
215.86
MFCD00067894
SULQOTGFPPFUMG-UHFFFAOYSA-N
2-bromo-4,5-dichloroimidazole, imidazole, 2-bromo-4,5-dichloro, 1h-imidazole, 2-bromo-4,5-dichloro, imidazole,5-dichloro, acmc-20ao2s, 1h-imidazole,5-dichloro, 5-23-04-00463 beilstein handbook reference, 2-bromo-4,5-dichloro-imidazole, 2-bromo-4,5-dichloro-3h-imidazole
99484
2-Brom-4,5-Dichlor-1H-Imidazol
ClC1=C(Cl)N=C(Br)N1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 2-Bromo-4,5-dichloroimidazole
Schmelzpunkt ∼215°C (decomposition)
Geruch Odorless
Menge 25 g
Beilstein 606732
Formelmasse 215.87
Reinheit (%) 98%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'