Learn More
2-Brom-5-methyl-4-phenylthiazol, 97 %, Thermo Scientific™
Catalog Number p-7034540
Click to view available options
Menge:
1 g
5 g
412923-45-6
C10H8BrNS
254.145
MFCD11220230
RNOQQISGUQUJMQ-UHFFFAOYSA-N
2-bromo-5-methyl-4-phenylthiazole, thiazole, 2-bromo-5-methyl-4-phenyl, hydrochloride 1:1
49651940
2-Brom-5-Methyl-4-Phenyl-1,3-Thiazol
CC1=C(N=C(S1)Br)C2=CC=CC=C2
412923-45-6
C10H8BrNS
254.145
MFCD11220230
RNOQQISGUQUJMQ-UHFFFAOYSA-N
2-bromo-5-methyl-4-phenylthiazole, thiazole, 2-bromo-5-methyl-4-phenyl, hydrochloride 1:1
49651940
2-Brom-5-Methyl-4-Phenyl-1,3-Thiazol
CC1=C(N=C(S1)Br)C2=CC=CC=C2
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Hinweise
Kühl lagern. Behälter an einem trockenen und gut belüfteten Ort dicht verschlossen halten. Von starken Oxidationsmitteln fernhalten.
TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 2-Bromo-5-methyl-4-phenylthiazole
Menge 1 g
Formelmasse 254.14
Reinheit (%) 97%