missing translation for 'onlineSavingsMsg'
Learn More

2-Brom-5-Methylbenzotrifluorid, 98 %, Thermo Scientific Chemicals

Catalog Number p-7030697
Click to view available options
Menge:
1 g
5 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
261952-20-9
C8H6BrF3
239.035
MFCD01631586
WXXDBPIOFOYRLP-UHFFFAOYSA-N
2-bromo-5-methylbenzotrifluoride, 1-bromo-4-methyl-2-trifluoromethyl benzene, 4-bromo-3-trifluoromethyl toluene, 4-bromo-alpha,alpha,alpha-trifluoro-m-xylene, 4-methyl-2-trifluoromethyl bromobenzene, pubchem23748, 4-bromo-3-trifluoromethyltoluene, 1-bromo-4-methyl-2-trifluoromethyl-benzene, 1-bromanyl-4-methyl-2-trifluoromethyl benzene, benzene,1-bromo-4-methyl-2-trifluoromethyl
2779339
1-Brom-4-methyl-2-(trifluormethyl)benzol
CC1=CC(=C(C=C1)Br)C(F)(F)F
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
261952-20-9
C8H6BrF3
239.035
MFCD01631586
WXXDBPIOFOYRLP-UHFFFAOYSA-N
2-bromo-5-methylbenzotrifluoride, 1-bromo-4-methyl-2-trifluoromethyl benzene, 4-bromo-3-trifluoromethyl toluene, 4-bromo-alpha,alpha,alpha-trifluoro-m-xylene, 4-methyl-2-trifluoromethyl bromobenzene, pubchem23748, 4-bromo-3-trifluoromethyltoluene, 1-bromo-4-methyl-2-trifluoromethyl-benzene, 1-bromanyl-4-methyl-2-trifluoromethyl benzene, benzene,1-bromo-4-methyl-2-trifluoromethyl
2779339
1-Brom-4-methyl-2-(trifluormethyl)benzol
CC1=CC(=C(C=C1)Br)C(F)(F)F

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 2-Bromo-5-methylbenzotrifluoride
Menge 1 g
Formelmasse 239.04
Reinheit (%) 98%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'