missing translation for 'onlineSavingsMsg'
Learn More

2-Brom-N-Methylbenzenesulfonamid, 97 %, Thermo Scientific™

Catalog Number p-7040457
Click to view available options
Menge:
250 mg
1 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
98192-14-4
C7H8BrNO2S
250.11
MFCD03084756
HSCKMNXETNFFAJ-UHFFFAOYSA-N
2-bromo-n-methylbenzenesulphonamide, 2-bromo-n-methylbenzene-1-sulfonamide, 2-bromo-n-methyl-benzenesulfonamide, acmc-209s9w, 2-bromo-n-methylbenzenesulfomide, n-methyl 2-bromobenzenesulphonamide, 2-bromanyl-n-methyl-benzenesulfonamide, benzenesulfonamide, 2-bromo-n-methyl
850141
2-bromo-N-methylbenzene-1-sulfonamide
CNS(=O)(=O)C1=CC=CC=C1Br
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
98192-14-4
C7H8BrNO2S
250.11
MFCD03084756
HSCKMNXETNFFAJ-UHFFFAOYSA-N
2-bromo-n-methylbenzenesulphonamide, 2-bromo-n-methylbenzene-1-sulfonamide, 2-bromo-n-methyl-benzenesulfonamide, acmc-209s9w, 2-bromo-n-methylbenzenesulfomide, n-methyl 2-bromobenzenesulphonamide, 2-bromanyl-n-methyl-benzenesulfonamide, benzenesulfonamide, 2-bromo-n-methyl
850141
2-bromo-N-methylbenzene-1-sulfonamide
CNS(=O)(=O)C1=CC=CC=C1Br

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 2-Bromo-N-methylbenzenesulfonamide
Menge 250 mg
Formelmasse 250.11
Reinheit (%) 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'