missing translation for 'onlineSavingsMsg'
Learn More

2-Brom-p-Xylol, 98+ %, Thermo Scientific Chemicals

Catalog Number p-7021848
Click to view available options
Menge:
25 g
100 g
500 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
553-94-6
C8H9Br
185.06
MFCD00000074
QXISTPDUYKNPLU-UHFFFAOYSA-N
2,5-dimethylbromobenzene, 2-bromo-p-xylene, 1-bromo-2,5-dimethylbenzene, benzene, 2-bromo-1,4-dimethyl, bromo-p-xylene, 2-bromo-1,4-dimethyl-benzene, 2,5-xylyl bromide, 2-bromo-1,4-xylene, 2,5-dimethylphenyl bromide, p-xylene, 2-bromo
11121
2-Brom-1,4-Dimethylbenzol
CC1=CC=C(C)C(Br)=C1
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
553-94-6
C8H9Br
185.06
MFCD00000074
QXISTPDUYKNPLU-UHFFFAOYSA-N
2,5-dimethylbromobenzene, 2-bromo-p-xylene, 1-bromo-2,5-dimethylbenzene, benzene, 2-bromo-1,4-dimethyl, bromo-p-xylene, 2-bromo-1,4-dimethyl-benzene, 2,5-xylyl bromide, 2-bromo-1,4-xylene, 2,5-dimethylphenyl bromide, p-xylene, 2-bromo
11121
2-Brom-1,4-Dimethylbenzol
CC1=CC=C(C)C(Br)=C1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 2-Bromo-p-xylene
Schmelzpunkt 9°C to 10°C
Dichte 1.347
Siedepunkt 199°C to 201°C
Menge 25 g
Brechungsindex 1.551
Beilstein 2040350
Formelmasse 185.07
Reinheit (%) ≥98%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'