Learn More
2-(Bromomethyl)-1-fluoro-3-(trifluoromethyl)benzene, 98%, BLD Pharm™
Product Code. 30947470
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
25 g
Unit Size:
1g
250mg
25g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30947470 1 g 1g
30942311 25 g 25g
30946915 5 g 5g
30935386 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 30947470 Supplier BLD Pharm Supplier No. BD108621g
 Restricted Item
Verfügbar ab: 27-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Specifications
CAS 239087-08-2
Molecular Formula C8H5BrF4
MDL Number MFCD00082477
Quantity 1 g
SMILES FC(C1=C(CBr)C(F)=CC=C1)(F)F
Molecular Weight (g/mol) 257.02
Percent Purity 98%
Chemical Name or Material 2-(Bromomethyl)-1-fluoro-3-(trifluoromethyl)benzene