missing translation for 'onlineSavingsMsg'
Learn More

2-Bromovalersäure, 98 %

Catalog Number 15498859
Click to view available options
Menge:
25 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 2-Bromovaleric acid
CAS 584-93-0
Dichte 1.381
Siedepunkt 132°C to 136°C (25 mmHg)
Flammpunkt >110°C (230°F)
Summenformel C5H9BrO2
Brechungsindex 1.471
MDL-Nummer MFCD00004217
Menge 25 g
UN-Nummer UN3265
Beilstein 1721172
Synonym 2-bromovaleric acid, pentanoic acid, 2-bromo, .alpha.-bromovaleric acid, valeric acid, 2-bromo, alpha-bromo-n-valeric acid, .alpha.-bromopentanoic acid, valeric acid, .alpha.-bromo, alpha-bromovaleric acid, alpha-bromopentanoic acid, valeric acid, alpha-bromo
InChI-Schlüssel WMFATTFQNRPXBQ-UHFFFAOYNA-N
SMILES CCCC(Br)C(O)=O
IUPAC-Name 2-bromopentanoic acid
Molekulargewicht (g/mol) 181.03
PubChem CID 11446
Formelmasse 181.03
Reinheit (%) 98%
Mehr anzeigen Weniger anzeigen
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'