Learn More
2-Carbamoyl-N-methylpyridinium Methanesulfonate, Mikromol™
Product Code. 17962566
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17962566 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17962566 Supplier Mikromol Supplier No. MM0689.020025
 Restricted Item
Verfügbar ab: 30-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2-Carbamoyl-N-methylpyridinium Methanesulfonate
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 2-Carbamoyl-N-methylpyridinium Methanesulfonate
CAS 87141-87-5
Analyte or Component Names 2-Carbamoyl-N-methylpyridinium Methanesulfonate
Shipping Condition Room Temperature
Molecular Formula C7 H9 N2 O . C H3 O3 S
Certifications/Compliance ISO 9001
Synonym 2-Carbamoyl-N-methylpyridinium Methanesulfonate
InChI Formula InChI=1S/C7H8N2O.CH4O3S/c1-9-5-3-2-4-6(9)7(8)10;1-5(2,3)4/h2-5H,1H3,(H-,8,10);1H3,(H,2,3,4)
SMILES C[n+]1ccccc1C(=O)N.CS(=O)(=O)[O-]
IUPAC Name methanesulfonate;1-methylpyridin-1-ium-2-carboxamide
Molecular Weight (g/mol) 232.26
Quantity 25 mg
Formula Weight 232.0518 g/mol