missing translation for 'onlineSavingsMsg'
Learn More

2-Chlor-2'-fluor-3,3'-bipyridin, 95 %, Thermo Scientific™

Catalog Number p-7029425
Click to view available options
Menge:
100 mg
500 mg
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
942206-09-9
C10H6ClFN2
208.62
MFCD09037427
XJKOECSGYQHCPP-UHFFFAOYSA-N
2-chloro-2'-fluoro-3,3'-bipyridine, 2-chloro-2'-fluoro-3,3' bipyridine, 3-2-chloropyridin-3-yl-2-fluoropyridine
42552970
2-Chlor-3-(2-Fluorpyridin-3-yl)Pyridin
FC1=C(C=CC=N1)C1=C(Cl)N=CC=C1
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
942206-09-9
C10H6ClFN2
208.62
MFCD09037427
XJKOECSGYQHCPP-UHFFFAOYSA-N
2-chloro-2'-fluoro-3,3'-bipyridine, 2-chloro-2'-fluoro-3,3' bipyridine, 3-2-chloropyridin-3-yl-2-fluoropyridine
42552970
2-Chlor-3-(2-Fluorpyridin-3-yl)Pyridin
FC1=C(C=CC=N1)C1=C(Cl)N=CC=C1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 2-Chloro-2'-fluoro-3,3'-bipyridine
Menge 100 mg
Formelmasse 208.62
Reinheit (%) 95%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'