missing translation for 'onlineSavingsMsg'
Learn More

2-Chlor-4‚-Fluoroacetanilid, 98 %, Thermo Scientific™

Catalog Number p-7023779
Click to view available options
Menge:
5 g
25 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
351-04-2
C8H7ClFNO
187.60
MFCD00015730
JDAWWCJBFPBHFL-UHFFFAOYSA-N
2-chloro-n-4-fluorophenyl acetamide, n-chloroacetyl-4-fluoroaniline, 2-chloro-4'-fluoroacetanilide, alpha-chloro-4-fluoroacetanilide, 2-chloro-n-4-fluoro-phenyl-acetamide, acetamide, n-4-fluorophenyl-2-chloro, acmc-20akao, pubchem8401, 4'-fluoro-2-chloroacetoanilide
532065
2-Chlor-N-(4-fluorphenyl)acetamid
FC1=CC=C(NC(=O)CCl)C=C1
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
351-04-2
C8H7ClFNO
187.60
MFCD00015730
JDAWWCJBFPBHFL-UHFFFAOYSA-N
2-chloro-n-4-fluorophenyl acetamide, n-chloroacetyl-4-fluoroaniline, 2-chloro-4'-fluoroacetanilide, alpha-chloro-4-fluoroacetanilide, 2-chloro-n-4-fluoro-phenyl-acetamide, acetamide, n-4-fluorophenyl-2-chloro, acmc-20akao, pubchem8401, 4'-fluoro-2-chloroacetoanilide
532065
2-Chlor-N-(4-fluorphenyl)acetamid
FC1=CC=C(NC(=O)CCl)C=C1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 2-Chloro-4'-fluoroacetanilide
Menge 5 g
Formelmasse 187.6
Reinheit (%) 98%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'