missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4-fluorocinnamic acid, 97%, Thermo Scientific™

Product Code. 11406256
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11406256

Brand: Thermo Scientific Alfa Aesar B23062.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 618 41 11 for assistance
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 133220-86-7
Molecular Formula C9H6ClFO2
Molecular Weight (g/mol) 200.593
MDL Number MFCD00236027
InChI Key RJCWBTRMWGOREZ-DUXPYHPUSA-N
Synonym 2-chloro-4-fluorocinnamic acid, rarechem bk hw 0063, timtec-bb sbb003541, 3-2-chloro-4-fluorophenyl-2-propenoic acid, 2e-3-2-chloro-4-fluorophenyl prop-2-enoic acid, 3-2-chloro-4-fluorophenyl acrylic acid, pubchem3429, e-3-2-chloro-4-fluorophenyl acrylic acid, 3-chloro-5-fluorobenzoic, 2-chloro-4-fluorocinnamicacid
PubChem CID 6257297
IUPAC Name (E)-3-(2-chloro-4-fluorophenyl)prop-2-enoic acid
SMILES C1=CC(=C(C=C1F)Cl)C=CC(=O)O

Specifications

Melting Point 245°C to 249°C
Quantity 5 g
Formula Weight 200.6
Percent Purity 97%
Chemical Name or Material 2-Chloro-4-fluorocinnamic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.