Learn More
2-Chlor-5-(Dimethylcarbamoyl)benzenboronsäure, 97 %
Catalog Number p-7036373
Click to view available options
Menge:
250 mg
1 g
1449144-75-5
C9H11BClNO3
227.451
MFCD22125080
LVOHUMGEBQAIBQ-UHFFFAOYSA-N
2-Chloro-5-(dimethylcarbamoyl)phenylboronic acid
99738090
[2-chlor-5-(dimethylcarbamoyl)phenyl]boronsäure
B(C1=C(C=CC(=C1)C(=O)N(C)C)Cl)(O)O
1449144-75-5
C9H11BClNO3
227.451
MFCD22125080
LVOHUMGEBQAIBQ-UHFFFAOYSA-N
2-Chloro-5-(dimethylcarbamoyl)phenylboronic acid
99738090
[2-chlor-5-(dimethylcarbamoyl)phenyl]boronsäure
B(C1=C(C=CC(=C1)C(=O)N(C)C)Cl)(O)O
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 2-Chloro-5-(dimethylcarbamoyl)benzeneboronic acid
Menge 250 mg
Formelmasse 227.45
Reinheit (%) 97%