missing translation for 'onlineSavingsMsg'
Learn More

2-Chlor-5-(Pentafluorothio)Benzamid, 97 %

Catalog Number p-7032964
Click to view available options
Menge:
250 mg
1 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
1431329-71-3
C7H5ClF5NOS
281.625
MFCD22988934
ISPQOXNUNOTXHB-UHFFFAOYSA-N
2-chloro-5-pentafluorothio benzamide, 2-chloro-5-pentafluorosulfur benzamide, 2-chloro-5-pentafluoro-??-sulfanyl benzamide
73995213
2-chlor-5-(pentafluor-$l^{6}-sulfanyl)benzamid
C1=CC(=C(C=C1S(F)(F)(F)(F)F)C(=O)N)Cl
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
1431329-71-3
C7H5ClF5NOS
281.625
MFCD22988934
ISPQOXNUNOTXHB-UHFFFAOYSA-N
2-chloro-5-pentafluorothio benzamide, 2-chloro-5-pentafluorosulfur benzamide, 2-chloro-5-pentafluoro-??-sulfanyl benzamide
73995213
2-chlor-5-(pentafluor-$l^{6}-sulfanyl)benzamid
C1=CC(=C(C=C1S(F)(F)(F)(F)F)C(=O)N)Cl

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 2-Chloro-5-(pentafluorothio)benzamide
Menge 250 mg
Formelmasse 281.63
Reinheit (%) 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'