missing translation for 'onlineSavingsMsg'
Learn More

2-Chlor-5-(pentafluorthio)Benzoesäure, 97 %

Catalog Number p-7032932
Click to view available options
Menge:
250 mg
1 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
1431329-70-2
C7H4ClF5O2S
282.609
MFCD22988931
KNKMPGVWOPIPMH-UHFFFAOYSA-N
2-chloro-5-pentafluorothio benzoic acid, 2-chloro-5-pentafluorosulfur benzoic acid, 2-chloro-5-pentafluoro-??-sulfanyl benzoic acid
73995201
2-chlor-5-(pentafluor-$l^{6}-sulfanyl)benzoesäure
C1=CC(=C(C=C1S(F)(F)(F)(F)F)C(=O)O)Cl
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
1431329-70-2
C7H4ClF5O2S
282.609
MFCD22988931
KNKMPGVWOPIPMH-UHFFFAOYSA-N
2-chloro-5-pentafluorothio benzoic acid, 2-chloro-5-pentafluorosulfur benzoic acid, 2-chloro-5-pentafluoro-??-sulfanyl benzoic acid
73995201
2-chlor-5-(pentafluor-$l^{6}-sulfanyl)benzoesäure
C1=CC(=C(C=C1S(F)(F)(F)(F)F)C(=O)O)Cl

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 2-Chloro-5-(pentafluorothio)benzoic acid
Menge 250 mg
Formelmasse 282.61
Reinheit (%) 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'