missing translation for 'onlineSavingsMsg'
Learn More

2-Chlor-6-Fluorbenzothiazol, 98 %, Thermo Scientific Chemicals

Catalog Number p-7031342
Click to view available options
Menge:
1 g
5 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
399-74-6
C7H3ClFNS
187.616
MFCD04448822
ISIIQFDYFMPPOA-UHFFFAOYSA-N
2-chloro-6-fluorobenzo d thiazole, 2-chloro-6-fluorobenzothiazole, 2-chloro-6-fluoro-benzothiazole, benzothiazole, 2-chloro-6-fluoro, pubchem21812, acmc-1aihk, ksc497m8n, 2-chloro-6-fluoro benzothiazole, benzothiazole, 2-chloro-6-fluoro-8ci,9ci
2049870
2-Chlor-6-Fluoro-1,3-Benzothiazol
C1=CC2=C(C=C1F)SC(=N2)Cl
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
399-74-6
C7H3ClFNS
187.616
MFCD04448822
ISIIQFDYFMPPOA-UHFFFAOYSA-N
2-chloro-6-fluorobenzo d thiazole, 2-chloro-6-fluorobenzothiazole, 2-chloro-6-fluoro-benzothiazole, benzothiazole, 2-chloro-6-fluoro, pubchem21812, acmc-1aihk, ksc497m8n, 2-chloro-6-fluoro benzothiazole, benzothiazole, 2-chloro-6-fluoro-8ci,9ci
2049870
2-Chlor-6-Fluoro-1,3-Benzothiazol
C1=CC2=C(C=C1F)SC(=N2)Cl

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 2-Chloro-6-fluorobenzothiazole
Schmelzpunkt 89°C to 93°C
Menge 1 g
Formelmasse 187.62
Reinheit (%) 98%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'