missing translation for 'onlineSavingsMsg'
Learn More

2-Chlor-N-(2,6-dichlorphenyl)benzamid, 97 %, Thermo Scientific™

Catalog Number p-7039964
Click to view available options
Menge:
250 mg
1 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
200807-45-0
C13H8Cl3NO
300.563
MFCD00028398
MLILWLKTWNAOFL-UHFFFAOYSA-N
2-chloro-n-2,6-dichlorophenyl benzamide, 2,2',6'-trichlorobenzanilide
853837
2-Chlor-N-(2,6-dichlorphenyl)benzamid
C1=CC=C(C(=C1)C(=O)NC2=C(C=CC=C2Cl)Cl)Cl
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
200807-45-0
C13H8Cl3NO
300.563
MFCD00028398
MLILWLKTWNAOFL-UHFFFAOYSA-N
2-chloro-n-2,6-dichlorophenyl benzamide, 2,2',6'-trichlorobenzanilide
853837
2-Chlor-N-(2,6-dichlorphenyl)benzamid
C1=CC=C(C(=C1)C(=O)NC2=C(C=CC=C2Cl)Cl)Cl

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 2-Chloro-N-(2,6-dichlorophenyl)benzamide
Menge 250 mg
Formelmasse 300.57
Reinheit (%) 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'