missing translation for 'onlineSavingsMsg'
Learn More

2-Chlor-N-(3-cyano-5,6-dihydro-4H-cyclopenta[b]thiophen-2 -yl)acetamid, 96 %, Alfa Aesar™

Catalog Number p-7032465
Click to view available options
Menge:
1 g
5 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
304685-87-8
C10H9ClN2OS
240.705
MFCD00454946
IFPYPHZLEFBRSS-UHFFFAOYSA-N
3568544
2-Chlor-N-(3-cyano-5,6-dihydro-4H-cyclopenta[b]thiophen-2-yl)acetamid
C1CC2=C(C1)SC(=C2C#N)NC(=O)CCl
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
304685-87-8
C10H9ClN2OS
240.705
MFCD00454946
IFPYPHZLEFBRSS-UHFFFAOYSA-N
3568544
2-Chlor-N-(3-cyano-5,6-dihydro-4H-cyclopenta[b]thiophen-2-yl)acetamid
C1CC2=C(C1)SC(=C2C#N)NC(=O)CCl

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 2-Chloro-N-(3-cyano-5,6-dihydro-4H-cyclopenta[b]thiophen-2-yl)acetamide
Schmelzpunkt 201°C to 202°C
Menge 1 g
Synonym 2-chloro-n-3-cyano-5,6-dihydro-4h-cyclopenta b thiophen-2-yl-acetamide, 2-chloro-n-3-cyano-5,6-dihydro-4h-cyclopenta b thiophen-2-yl acetamide, 2-chloro-n-3-cyano-4h,5h,6h-cyclopenta b thiophen-2-yl acetamide, 2-chloro-n-3-cyano-5,6-dihydro-4h-cyclopenta b thien-2-yl acetamide, acetamide, 2-chloro-n-3-cyano-5,6-dihydro-4h-cyclopenta b thien-2-yl, n-3-cyano-5,6-dihydro-4h-cyclopenta b thiophene-2-yl chloroacetamide, 2-chloro-n-3-cyano 4,5,6-trihydrocyclopenta 1,2-d thiophen-2-yl acetamide, acetamide,2-chloro-n-3-cyano-5,6-dihydro-4h-cyclopenta b thien-2-yl
Formelmasse 240.71
Reinheit (%) 96%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'