missing translation for 'onlineSavingsMsg'
Learn More

2-Chlor-N-Cyclopropylacetamid, 97 %, Thermo Scientific Chemicals

Catalog Number 15455177
Click to view available options
Menge:
10 g
50 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
19047-31-5
C5H8ClNO
133.58
MFCD00276391
ZPWIVSGEQGESFF-UHFFFAOYSA-N
n1-cyclopropyl-2-chloroacetamide, 2-chloro-n-cyclopropyl-acetamide, n-cyclopropylchloroacetamide, acetamide,2-chloro-n-cyclopropyl
735780
2-chloro-N-cyclopropylacetamide
ClCC(=O)NC1CC1
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
19047-31-5
C5H8ClNO
133.58
MFCD00276391
ZPWIVSGEQGESFF-UHFFFAOYSA-N
n1-cyclopropyl-2-chloroacetamide, 2-chloro-n-cyclopropyl-acetamide, n-cyclopropylchloroacetamide, acetamide,2-chloro-n-cyclopropyl
735780
2-chloro-N-cyclopropylacetamide
ClCC(=O)NC1CC1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 2-Chloro-N-cyclopropylacetamide
Schmelzpunkt 80°C to 84°C
Menge 50 g
Formelmasse 133.58
Reinheit (%) 97%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'