missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorpyridin-4-carbonsäure, 98 %, Thermo Scientific Chemicals

Catalog Number p-7047562
Click to view available options
Menge:
1 g
5 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
6313-54-8
C6H4ClNO2
157.55
MFCD00191402
QXCOHSRHFCHCHN-UHFFFAOYSA-N
2-chloroisonicotinic acid, 2-chloro-4-pyridinecarboxylic acid, 2-chloro-isonicotinic acid, 2-chloro isonicotinic acid, 4-pyridinecarboxylic acid, 2-chloro, 2-chloro-4-pyridinyl carboxylic acid, 2-chlorisonicotins, pubchem2598, 2-chloroisonicotinic acid,, 4-carboxy-2-chloropyridine
236985
2-Chlorpyridin-4-Carbonsäure
OC(=O)C1=CC=NC(Cl)=C1
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
6313-54-8
C6H4ClNO2
157.55
MFCD00191402
QXCOHSRHFCHCHN-UHFFFAOYSA-N
2-chloroisonicotinic acid, 2-chloro-4-pyridinecarboxylic acid, 2-chloro-isonicotinic acid, 2-chloro isonicotinic acid, 4-pyridinecarboxylic acid, 2-chloro, 2-chloro-4-pyridinyl carboxylic acid, 2-chlorisonicotins, pubchem2598, 2-chloroisonicotinic acid,, 4-carboxy-2-chloropyridine
236985
2-Chlorpyridin-4-Carbonsäure
OC(=O)C1=CC=NC(Cl)=C1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 2-Chloropyridine-4-carboxylic acid
Schmelzpunkt 230°C to 232°C
Menge 1 g
Beilstein 115861
Formelmasse 157.56
Reinheit (%) 98%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'