Learn More
(2-Cyanoethyl)ethylcarbaminsäure-tert-butylester, TRC
Specifications
Specifications
| Chemischer Name oder Material | (2-Cyanoethyl)ethyl-carbamic Acid tert-Butyl Ester |
| CAS | 170018-97-0 |
| Summenformel | C10H18N2O2 |
| Menge | 2.5 g |
| Synonym | (2-Cyanoethyl)ethyl-carbamic Acid 1,1-Dimethylethyl Ester, tert-Butyl N-(2-Cyanoethyl)-N-(ethyl)carbamate |
| InChI-Formel | InChI=1S/C10H18N2O2/c1-5-12(8-6-7-11)9(13)14-10(2,3)4/h5-6,8H2,1-4H3 |
| SMILES | CCN(CCC#N)C(=O)OC(C)(C)C |
| IUPAC-Name | tert-Butyl-N-(2-cyanoethyl)-N-ethylcarbamat |
| Molekulargewicht (g/mol) | 198.26 |
| Formelmasse | 198.1368 |