Learn More
Alfa Aesar™ 2-Ethoxypyridin-3-borsäure, 98 %
Marke: Alfa Aesar™ L20422.03
Weitere Details : CAS : 854373-97-0 Netto-Gewicht : 0.00100kg
Menge | 1g |
---|
Chemische Identifikatoren
854373-97-0 | |
166.971 | |
FXUMKSCYKPOZOO-UHFFFAOYSA-N | |
11309733 | |
B(C1=C(N=CC=C1)OCC)(O)O |
C7H10BNO3 | |
MFCD06201036 | |
2-ethoxypyridine-3-boronic acid, 2-ethoxy-3-pyridineboronic acid, 2-ethoxypyridin-3-yl boronic acid, 2-ethoxy-3-pyridyl boronic acid, 3-borono-2-ethoxypyridine, pubchem5119, acmc-209q5z, 2-ethoxypyridine-3-boronicacid, 2-ethoxy-3-pyridinyl boronic acid, 2-ethoxy-pyridin-3-yl-boronic acid | |
(2-Ethoxypyridin-3-yl)borsäure |
Spezifikation
2-Ethoxypyridine-3-boronic acid | |
854373-97-0 | |
MFCD06201036 | |
FXUMKSCYKPOZOO-UHFFFAOYSA-N | |
(2-Ethoxypyridin-3-yl)borsäure | |
11309733 | |
98% |
1g | |
C7H10BNO3 | |
2-ethoxypyridine-3-boronic acid, 2-ethoxy-3-pyridineboronic acid, 2-ethoxypyridin-3-yl boronic acid, 2-ethoxy-3-pyridyl boronic acid, 3-borono-2-ethoxypyridine, pubchem5119, acmc-209q5z, 2-ethoxypyridine-3-boronicacid, 2-ethoxy-3-pyridinyl boronic acid, 2-ethoxy-pyridin-3-yl-boronic acid | |
B(C1=C(N=CC=C1)OCC)(O)O | |
166.971 | |
166.97 |