missing translation for 'onlineSavingsMsg'
Learn More

2-Ethylthio-1,3,4-Thiadiazol-5-Thiol, 97 %, Thermo Scientific Chemicals

Catalog Number p-7026460
Click to view available options
Menge:
1 g
5 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
37147-15-2
C4H6N2S3
178.29
MFCD00186131
JLJYQXMUUKOBBO-UHFFFAOYSA-N
2-ethylthio-1,3,4-thiadiazole-5-thiol, 5-ethylthio-1,3,4-thiadiazole-2-thiol, 5-ethylsulfanyl-1,3,4-thiadiazole-2-thiol, 2-ethylthio-1,3,4-thiazole-5-thiol, 5-ethylthio-1,3,4-thiadiazole-2 3h-thione, 5-ethylsulfanyl-1,3,4-thiadiazole-2 3h-thione, acmc-1cszq, 5-ethylthio-3h-1,3,4-thiadiazole-2-thione
1201366
5-Ethylsulfanyl-3H-1,3,4-thiadiazol-2-thion
CCSC1=NNC(=S)S1
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
37147-15-2
C4H6N2S3
178.29
MFCD00186131
JLJYQXMUUKOBBO-UHFFFAOYSA-N
2-ethylthio-1,3,4-thiadiazole-5-thiol, 5-ethylthio-1,3,4-thiadiazole-2-thiol, 5-ethylsulfanyl-1,3,4-thiadiazole-2-thiol, 2-ethylthio-1,3,4-thiazole-5-thiol, 5-ethylthio-1,3,4-thiadiazole-2 3h-thione, 5-ethylsulfanyl-1,3,4-thiadiazole-2 3h-thione, acmc-1cszq, 5-ethylthio-3h-1,3,4-thiadiazole-2-thione
1201366
5-Ethylsulfanyl-3H-1,3,4-thiadiazol-2-thion
CCSC1=NNC(=S)S1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 2-Ethylthio-1,3,4-thiadiazole-5-thiol
Schmelzpunkt 119°C to 122°C
Menge 1 g
Formelmasse 178.3
Reinheit (%) 97%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'