missing translation for 'onlineSavingsMsg'
Learn More

2-Fluor-1,4-bis-(trifluormethyl)-benzol, 95 %, Thermo Scientific™

Catalog Number p-7031957
Click to view available options
Menge:
250 mg
1 g
5 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
887268-08-8
C8H3F7
232.10
MFCD03412236
KAGRJEBZBWYSJK-UHFFFAOYSA-N
2,5-bis trifluoromethyl fluorobenzene, 2-fluoro-1,4-bis-trifluoromethyl benzene, 1,4-bis trifluoromethyl-2-fluorobenzene, 2-fluoro-1,4-bis trifluoromethyl benzene
44717336
2-fluoro-1,4-bis(trifluoromethyl)benzene
FC1=C(C=CC(=C1)C(F)(F)F)C(F)(F)F
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
887268-08-8
C8H3F7
232.10
MFCD03412236
KAGRJEBZBWYSJK-UHFFFAOYSA-N
2,5-bis trifluoromethyl fluorobenzene, 2-fluoro-1,4-bis-trifluoromethyl benzene, 1,4-bis trifluoromethyl-2-fluorobenzene, 2-fluoro-1,4-bis trifluoromethyl benzene
44717336
2-fluoro-1,4-bis(trifluoromethyl)benzene
FC1=C(C=CC(=C1)C(F)(F)F)C(F)(F)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 2-Fluoro-1,4-bis(trifluoromethyl)benzene
Siedepunkt 50°C to 54°C (60 mmHg)
Menge 250 mg
Formelmasse 232.09
Reinheit (%) 95%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'