missing translation for 'onlineSavingsMsg'
Learn More

2-Fluor-4-(Methylcarbamoyl)benzenboronsäure, 97 %, Thermo Scientific™

Catalog Number p-7037116
Click to view available options
Menge:
250 mg
1 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
874289-23-3
C8H9BFNO3
196.972
MFCD11856043
STYRVOIKGOQNAD-UHFFFAOYSA-N
2-fluoro-4-methylcarbamoyl phenyl boronic acid, 2-fluoro-4-methylcarbamoyl benzeneboronic acid, 4-n-methylaminocarbonyl-2-fluorophenylboronic acid, 2-fluoro-4-methylcarbamoyl phenylboronic acid
57497292
[2-fluor-4-(methylcarbamoyl)phenyl]boronsäure
B(C1=C(C=C(C=C1)C(=O)NC)F)(O)O
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
874289-23-3
C8H9BFNO3
196.972
MFCD11856043
STYRVOIKGOQNAD-UHFFFAOYSA-N
2-fluoro-4-methylcarbamoyl phenyl boronic acid, 2-fluoro-4-methylcarbamoyl benzeneboronic acid, 4-n-methylaminocarbonyl-2-fluorophenylboronic acid, 2-fluoro-4-methylcarbamoyl phenylboronic acid
57497292
[2-fluor-4-(methylcarbamoyl)phenyl]boronsäure
B(C1=C(C=C(C=C1)C(=O)NC)F)(O)O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 2-Fluoro-4-(methylcarbamoyl)benzeneboronic acid
Menge 250 mg
Formelmasse 196.97
Reinheit (%) 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'