missing translation for 'onlineSavingsMsg'
Learn More

2-Fluor-5-(2-fluor-5-nitrophenylcarbamoyl)benzenboronsäure, 97 %, Thermo Scientific™

Catalog Number p-7036474
Click to view available options
Menge:
250 mg
1 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
1449133-65-6
C13H9BF2N2O5
322.03
MFCD20091227
MTQQOSSMDAQKRY-UHFFFAOYSA-N
2-fluoro-5-2-fluoro-5-nitrophenylcarbamoyl benzeneboronic acid, 2-fluoro-5-2-fluoro-5-nitrophenyl carbamoyl phenylboronic acid
73995958
{2-fluoro-5-[(2-fluoro-5-nitrophenyl)carbamoyl]phenyl}boronic acid
OB(O)C1=C(F)C=CC(=C1)C(=O)NC1=C(F)C=CC(=C1)[N+]([O-])=O
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
1449133-65-6
C13H9BF2N2O5
322.03
MFCD20091227
MTQQOSSMDAQKRY-UHFFFAOYSA-N
2-fluoro-5-2-fluoro-5-nitrophenylcarbamoyl benzeneboronic acid, 2-fluoro-5-2-fluoro-5-nitrophenyl carbamoyl phenylboronic acid
73995958
{2-fluoro-5-[(2-fluoro-5-nitrophenyl)carbamoyl]phenyl}boronic acid
OB(O)C1=C(F)C=CC(=C1)C(=O)NC1=C(F)C=CC(=C1)[N+]([O-])=O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 2-Fluoro-5-(2-fluoro-5-nitrophenylcarbamoyl)benzeneboronic acid
Menge 250 mg
Formelmasse 322.03
Reinheit (%) 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'