missing translation for 'onlineSavingsMsg'
Learn More

2-Fluor-5-Methyl-4-(Trifluormethyl)Benzylamin, 97 %, Thermo Scientific™

Catalog Number p-7032078
Click to view available options
Menge:
250 mg
1 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
1323966-36-4
C9H9F4N
207.172
MFCD19687196
RVXWFCFVRIODHK-UHFFFAOYSA-N
2-fluoro-5-methyl-4-trifluoromethyl benzylamine, 1-2-fluoro-5-methyl-4-trifluoromethyl phenyl methanamine, 2-fluoro-5-methyl-4-trifluoromethyl phenyl methanamine
66523485
[2-fluor-5-methyl-4-(trifluormethyl)phenyl]methanamin
CC1=CC(=C(C=C1C(F)(F)F)F)CN
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
1323966-36-4
C9H9F4N
207.172
MFCD19687196
RVXWFCFVRIODHK-UHFFFAOYSA-N
2-fluoro-5-methyl-4-trifluoromethyl benzylamine, 1-2-fluoro-5-methyl-4-trifluoromethyl phenyl methanamine, 2-fluoro-5-methyl-4-trifluoromethyl phenyl methanamine
66523485
[2-fluor-5-methyl-4-(trifluormethyl)phenyl]methanamin
CC1=CC(=C(C=C1C(F)(F)F)F)CN

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 2-Fluoro-5-methyl-4-(trifluoromethyl)benzylamine
Menge 250 mg
UN-Nummer UN3259
Empfindlichkeit Air sensitive
Formelmasse 207.17
Reinheit (%) 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'