Learn More
2-Fluoro-5-(trifluoromethyl)pyridine, 97%, Thermo Scientific™
Product Code. 11334656
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11334656 1 g 1g
11324656 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11334656 Supplier Thermo Scientific Alfa Aesar Supplier No. H32574.03
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 69045-82-5
Molecular Formula C6H3F4N
Molecular Weight (g/mol) 165.091
MDL Number MFCD07437945
InChI Key UUODQIKUTGWMPT-UHFFFAOYSA-N
Synonym 2-fluoro-5-trifluoromethyl pyridine, 2-fluoro-5-trifluoromethyl-pyridine, pyridine, 2-fluoro-5-trifluoromethyl, alpha,alpha,alpha,6-tetrafluoro-3-picoline, pubchem9252, acmc-209o5o, ksc628s4n
PubChem CID 5463754
IUPAC Name 2-fluoro-5-(trifluoromethyl)pyridine
SMILES C1=CC(=NC=C1C(F)(F)F)F
Specifications
Density 1.371
Boiling Point 118°C to 122°C
Flash Point 35°C (95°F)
Refractive Index 1.402
Quantity 1 g
UN Number UN1993
Formula Weight 165.09
Percent Purity 97%
Chemical Name or Material 2-Fluoro-5-(trifluoromethyl)pyridine