Learn More
2-Hydroxybutansäure-d3-Säure-Natriumsalz, TRC
Spezifikation
Spezifikation
| Chemischer Name oder Material | Sodium 2-Hydroxybutanoate-2,3,3-D3 |
| CAS | 1219798-97-6 |
| Summenformel | C4H4D3NaO3 |
| Menge | 10 mg |
| Synonym | (±)-2-Hydroxybutanoic-d3 Acid Sodium Salt, DL-2-Hydroxybutyric-d3 Acid Sodium Salt, (RS)-2-Hydroxybutyric-d3 Acid Sodium Salt, (±)-2-Hydroxy-n-butyric-d3 Acid Sodium Salt, (±)-2-Hydroxybutanoic-d3 Acid Sodium Salt, (±)-2-Hydroxybutyric-d3 Acid Sodium Salt, (±)-α-Hydroxybutyric-d3 Acid Sodium Salt, 2-Hydroxybutanoic-d3 Acid Sodium Salt, 2-Hydroxybutyric-d3 Acid Sodium Salt, DL-2-Hydroxybutanoic-d3 Acid Sodium Salt, DL-2-Hydroxybutyric-d3 Acid Sodium Salt, DL-α-Hydroxybutyric-d3 Acid Sodium Salt NSC 6495, α-Hydroxy-n-butyric-d3 Acid Sodium Salt, α-Hydroxybutanoic-d3 Acid Sodium Salt, α-Hydroxybutyric-d3 Acid Sodium Salt |
| InChI-Formel | InChI=1S/C4H8O3.Na/c1-2-3(5)4(6)7;/h3,5H,2H2,1H3,(H,6,7);/q;+1/p-1/i2D2,3D; |
| SMILES | OC(C(O[Na])=O)([2H])C([2H])([2H])C |
| IUPAC-Name | Natrium-2-Hydroxybutanoat-2,3,3-d3 |
| Molekulargewicht (g/mol) | 129.1 |
| Formelmasse | 129.05 |