missing translation for 'onlineSavingsMsg'
Learn More

2-Isobutyl-3-methoxypyrazin, 98 %, Thermo Scientific Chemicals

Catalog Number p-7021374
Click to view available options
Menge:
1 g
5 g
25 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
24683-00-9
C9H14N2O
166.22
MFCD00006128
UXFSPRAGHGMRSQ-UHFFFAOYSA-N
2-isobutyl-3-methoxypyrazine, 3-isobutyl-2-methoxypyrazine, 2-methoxy-3-2-methylpropyl pyrazine, 2-methoxy-3-isobutylpyrazine, 2-methoxy-3-isobutyl pyrazine, 1dzk, pyrazine, 2-methoxy-3-2-methylpropyl, unii-s327o0t12o, ibmp, 3-methoxy-2-isobutylpyrazine
32594
2-Methoxy-3-(2-Methylpropyl)Pyrazin
COC1=NC=CN=C1CC(C)C
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
24683-00-9
C9H14N2O
166.22
MFCD00006128
UXFSPRAGHGMRSQ-UHFFFAOYSA-N
2-isobutyl-3-methoxypyrazine, 3-isobutyl-2-methoxypyrazine, 2-methoxy-3-2-methylpropyl pyrazine, 2-methoxy-3-isobutylpyrazine, 2-methoxy-3-isobutyl pyrazine, 1dzk, pyrazine, 2-methoxy-3-2-methylpropyl, unii-s327o0t12o, ibmp, 3-methoxy-2-isobutylpyrazine
32594
2-Methoxy-3-(2-Methylpropyl)Pyrazin
COC1=NC=CN=C1CC(C)C

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 2-Isobutyl-3-methoxypyrazine
Dichte 0.986
Flammpunkt 80°C (176°F)
Brechungsindex 1.49
Menge 1 g
Formelmasse 166.22
Reinheit (%) 98%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'