missing translation for 'onlineSavingsMsg'
Learn More

2-Isobutyl-4,5-dimethyl-3-thiazolin, 97 %, Thermo Scientific™

Catalog Number p-7028960
Click to view available options
Menge:
1 g
10 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
65894-83-9
C9H17NS
171.30
MFCD01680822
FDOISHJOXPONIV-UHFFFAOYNA-N
4,5-dimethyl-2-isobutyl-3-thiazoline, 2-isobutyl-4,5-dimethyl-2,5-dihydrothiazole, 2-isobutyl-4,5-dimethyl-3-thiazoline, 2,5-dihydro-4,5-dimethyl-2-2-methylpropyl thiazole, thiazole, 2,5-dihydro-4,5-dimethyl-2-2-methylpropyl, fema no. 3621, 2-isobutyl-4,5-dimethyl thiazoline, 4,5-dimethyl-2-2-methylpropyl-2,5-dihydro-1,3-thiazole, 2,5-dihydro-2-isobutyl-4,5-dimethylthiazole, 3-thiazoline, 4,5-dimethyl-2-isobutyl
4120025
4,5-Dimethyl-2-(2-Methylpropyl)-2,5-Dihydro-1,3-Thiazol
CC(C)CC1SC(C)C(C)=N1
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
65894-83-9
C9H17NS
171.30
MFCD01680822
FDOISHJOXPONIV-UHFFFAOYNA-N
4,5-dimethyl-2-isobutyl-3-thiazoline, 2-isobutyl-4,5-dimethyl-2,5-dihydrothiazole, 2-isobutyl-4,5-dimethyl-3-thiazoline, 2,5-dihydro-4,5-dimethyl-2-2-methylpropyl thiazole, thiazole, 2,5-dihydro-4,5-dimethyl-2-2-methylpropyl, fema no. 3621, 2-isobutyl-4,5-dimethyl thiazoline, 4,5-dimethyl-2-2-methylpropyl-2,5-dihydro-1,3-thiazole, 2,5-dihydro-2-isobutyl-4,5-dimethylthiazole, 3-thiazoline, 4,5-dimethyl-2-isobutyl
4120025
4,5-Dimethyl-2-(2-Methylpropyl)-2,5-Dihydro-1,3-Thiazol
CC(C)CC1SC(C)C(C)=N1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 2-Isobutyl-4,5-dimethyl-3-thiazoline
Dichte 1.487
Siedepunkt 208°C
Flammpunkt 87°C (189°F)
Brechungsindex 1.487
Menge 1 g
Formelmasse 171.3
Reinheit (%) 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'