missing translation for 'onlineSavingsMsg'
Learn More

2-Isobutyl-4,5-dimethylthiazol, 99 %, Thermo Scientific™

Catalog Number p-7026523
Click to view available options
Menge:
2.5 g
10 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
53498-32-1
C9H15NS
169.29
MFCD00053136
NSVPHVLZAKJSGV-UHFFFAOYSA-N
2-isobutyl-4,5-dimethylthiazole, geranium thiazole, 4,5-dimethyl-2-isobutylthiazole, thiazole, 4,5-dimethyl-2-2-methylpropyl, 4,5-dimethyl-2-isobutyl thiazole, unii-5p82vd25zi, 4,5-dimethyl-2-2-methylpropyl-1,3-thiazole, acmc-1ak7t, 2-iso-butyl-4,5-dimethylthiazole, 2-isobutyl-4,5-dimethyl-thiazole
104513
4,5-Dimethyl-2-(2-Methylpropyl)-1,3-Thiazol
CC(C)CC1=NC(C)=C(C)S1
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
53498-32-1
C9H15NS
169.29
MFCD00053136
NSVPHVLZAKJSGV-UHFFFAOYSA-N
2-isobutyl-4,5-dimethylthiazole, geranium thiazole, 4,5-dimethyl-2-isobutylthiazole, thiazole, 4,5-dimethyl-2-2-methylpropyl, 4,5-dimethyl-2-isobutyl thiazole, unii-5p82vd25zi, 4,5-dimethyl-2-2-methylpropyl-1,3-thiazole, acmc-1ak7t, 2-iso-butyl-4,5-dimethylthiazole, 2-isobutyl-4,5-dimethyl-thiazole
104513
4,5-Dimethyl-2-(2-Methylpropyl)-1,3-Thiazol
CC(C)CC1=NC(C)=C(C)S1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 2-Isobutyl-4,5-dimethylthiazole
Dichte 0.967
Menge 2.5 g
Formelmasse 169.28
Reinheit (%) 99%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'